missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Binder™ Small Shelf Set for Environmental Walk-in Chambers
Shop All BINDER Products
Click to view available options
Unit Size:
Each
Description
- Accessory for WIC models 1 and 1 C
Specifications
Specifications
| Color | Silver |
| For Use With (Equipment) | Binder™ Environmental Walk-in Chambers 1 and 1 C |
| Material | Stainless Steel |
| Type | Shelf Set |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction