missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Small RNA Marker

Product Code. 16062038
Change view
Click to view available options
Quantity:
30 μL
Unit Size:
30µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16062038 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16062038 Supplier Abnova™ Supplier No. R0007.30uL
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Small RNAs include a variety of non-coding RNAs, such as miRNA, siRNA, snoRNA and snRNA

  • Recently such a small RNA is intensively studied, because the small RNA has been found to control many biological events.
  • The Small RNA Marker consists of five single-stranded RNAs (ssRNA).
  • The 20, 30, 40 and 50 bases RNAs are synthesized by chemically (non phosphorylated).
  • The 100 bases RNA is synthesized by in vitro transcription.
  • The Small RNA Marker is suitable for determinating size of small-size ssRNA in denaturing polyacrylamide gel electrophoresis.
  • The Small RNA Marker can be visualized by ethidium bromide staining.

Specifications

For Use With (Application) Electrophoresis
Quantity 30 μL
Storage Requirements Store at -80°C Repeated freeze/thaw cycles should be avoided
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.