missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Stainless Steel Small Hub Removable Custom Needles

Used for animal injections to limit tissue damage and sample loss.

Brand:  Hamilton™ 7731-10

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12336299

  • £320.00 / Pack of 6
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Autoclavable
  • Provided in packs of 6
  • Can be used with removable (RN)-style syringes that are 100μL and smaller
  • Custom lengths and point styles are available
  • Removable needles have minimal dead volume and for this reason are recommended over a luer or luer lock connection

Specifications

Specifications

Autoclavable
20 G
0.91 mm
6/Pk.
Stainless Steel
0.60 mm
Small Hub Removable Needle
5 or AS
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers