Learn More
Hamilton™ Stainless Steel Small Hub Removable Custom Needles
Product Code. 12311583
Change view
Click to view available options
Needle Point Style:
2, 3 or 4
2, 3, or 4
5 or AS
Unit Size:
Pack of 6
Needle Gauge:
20 G
25 G
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Point Style unitSize Needle Gauge
12311583 2, 3 or 4 Pack of 6 -
10509822 2, 3 or 4 Pack of 6 -
11512525 2, 3 or 4 Pack of 6 -
12396289 5 or AS Pack of 6 -
12376439 2, 3, or 4 Pack of 6 25 G
12326449 2, 3, or 4 Pack of 6 20 G
12336299 5 or AS Pack of 6 20 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12311583 Supplier Hamilton™ Supplier No. 780409
Add to basket
Add to basket
Used for animal injections to limit tissue damage and sample loss.
  • Autoclavable
  • Provided in packs of 6
  • Can be used with removable (RN)-style syringes that are 100μL and smaller
  • Custom lengths and point styles are available
  • Removable needles have minimal dead volume and for this reason are recommended over a luer or luer lock connection
Specifications
Needle Point Style 2, 3 or 4