Learn More
Hamilton™ Stainless Steel Small Hub Removable Custom Needles
Used for animal injections to limit tissue damage and sample loss.
Brand:  Hamilton™ 207419
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11512525
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Autoclavable
  • Provided in packs of 6
  • Can be used with removable (RN)-style syringes that are 100μL and smaller
  • Custom lengths and point styles are available
  • Removable needles have minimal dead volume and for this reason are recommended over a luer or luer lock connection
Specifications
Specifications
19 G
2, 3 or 4