Learn More
Heinz Herenz Small Foam Swabs Wooden stick
Product Code. 30158373
Click to view available options
Tip Type:
Minitip
Unit Size:
Pack of 4000
Product Code. 30158373
Brand: Heinz Herenz 1020010
Foam-swabs.
We offer a wide range of sterile and nonsterile swabs:
  • Wood, PS, PP, aluminium or paper applicator
  • Each in various lengths available
  • With cotton, viscose, polyester or foam head
  • Diameter of the heads: small, medium or large
  • This guarantees that the right instrument is available for every special application.
  • For example, the cotton-tipped swabs are recommended for use by ENT specialists.
  • Our sterile cotton buds are all individually wrapped.
Please note in the medical field:
  • Only suitable for single use
  • Use by medical personnel only
Specifications
Sterility Non-sterile
Applicator Material Polypropylene
Certifications/Compliance ISO 13485
Handle Material Wood
Shelf Life 5 Years
Tip Material Foam
Tip Type Minitip
Product Type Foam Swab