missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Small Core Pre-printed GHS CLP Labels

Product Code. 30041465
Change view
Click to view available options
Display Text:
1 Red Diamond per Sticker
2 Horizontal Red Diamonds per Sticker
2 Stacked Red Diamonds Plus 2 Additional Red Diamonds per Sticker
2 Stacked Red Diamonds Plus 2 Additional Red Diamons per Sticker
2 Stacked Red Diamonds per Sticker
3 Red Diamonds per Sticker
3 Red Diamonds per Sticker in Rotated Pyramidal Shape
3 Stacked Red Diamonds in Pyramid Formation per Sticker
4 Red Diamonds in Square Formation per Sticker
Width (Metric) Tape:
55 mm
55.00 mm
75.00 mm
105.00 mm
Material:
Paper
Polypropylene
Unit Size:
Pack of 200
Pack of 300
Pack of 400
Product Code. Display Text Width (Metric) Tape Material unitSize
30041465 3 Red Diamonds per Sticker in Rotated Pyramidal Shape 105.00 mm Polypropylene Pack of 200
30041449 1 Red Diamond per Sticker 55.00 mm Polypropylene Pack of 400
30041431 3 Red Diamonds per Sticker 75.00 mm Polypropylene Pack of 300
30041410 2 Stacked Red Diamonds per Sticker 105.00 mm Paper Pack of 200
30041461 2 Stacked Red Diamonds Plus 2 Additional Red Diamonds per Sticker 55.00 mm Polypropylene Pack of 400
30041411 2 Stacked Red Diamonds Plus 2 Additional Red Diamons per Sticker 55.00 mm Paper Pack of 400
30041412 3 Red Diamonds per Sticker 105.00 mm Paper Pack of 200
30041476 3 Stacked Red Diamonds in Pyramid Formation per Sticker 105.00 mm Paper Pack of 200
30041452 3 Stacked Red Diamonds in Pyramid Formation per Sticker 55 mm Paper Pack of 400
30041414 2 Stacked Red Diamonds per Sticker 55 mm Polypropylene Pack of 400
30041453 3 Red Diamonds per Sticker in Rotated Pyramidal Shape 55 mm Polypropylene Pack of 400
30041464 4 Red Diamonds in Square Formation per Sticker 55.00 mm Polypropylene Pack of 400
30041477 3 Red Diamonds per Sticker in Rotated Pyramidal Shape 75.00 mm Polypropylene Pack of 300
30041424 4 Red Diamonds in Square Formation per Sticker 105.00 mm Polypropylene Pack of 200
30041446 1 Red Diamond per Sticker 55.00 mm Paper Pack of 400
30041480 4 Red Diamonds in Square Formation per Sticker 75.00 mm Polypropylene Pack of 300
30041455 2 Horizontal Red Diamonds per Sticker 105.00 mm Polypropylene Pack of 200
30041482 1 Red Diamond per Sticker 105.00 mm Polypropylene Pack of 200
18 options
This item is not returnable. View return policy

Product Code. 30041465

Brand: Brady™ 622333

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Supplied with pre-printed red diamonds, ready for you to print the required warning, compliant to the latest regulations.

For you to print the required warning, compliant to the latest regulations.

Specifications

Certifications/Compliance UL Recognized
Description Pre-printed Desktop Printer Labels for CLP-GHS Hazardous Substances
Adhesive Type Acrylic, Permanent
Coating None
Color Red on White
Finish Gloss
Format Die-cut
Material Polypropylene
Printable Sides Front
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
Display Text 3 Red Diamonds per Sticker in Rotated Pyramidal Shape
For Use With (Application) Hazard communication identification
For Use With (Equipment) BBP11, BBP12, PR Plus, THT 25 mm Core Printers
Labels per Row 1
Min. Operating Temperature (Metric) -20°C
Model R6000HF
No. per Pack 200 Labels/Roll
Quantity 200 Stickers
Shape Rectangle
Size 105.00 mm (W) x 155.00 mm (H)
Temperature Range (English) -4°F to 212°F
Temperature Range (Metric) -20°C to +100°C
Thickness (English) 0.0031 in.
Thickness (Metric) 0.079 mm
Dimensions (LxW) 105.00 mm (W) x 155.00 mm (H)
Length (English) 6.1 in.
Length (Metric) 155.00 mm
Width (English) 4.13 in.
Width (Metric) 105.00 mm
Width (English) Printable Area 105.00 mm
Width (Metric) Tape 105.00 mm
Height (English) 6.1 in.
Height (Metric) 155.00 mm
Alternate Part Number THTCLP-L-L3A-7594-0.2-SC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.