missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Small Core Pre-printed GHS CLP Labels

Product Code. 30041477
Click to view available options
Display Text:
1 Red Diamond per Sticker
2 Horizontal Red Diamonds per Sticker
2 Stacked Red Diamonds Plus 2 Additional Red Diamonds per Sticker
2 Stacked Red Diamonds Plus 2 Additional Red Diamons per Sticker
2 Stacked Red Diamonds per Sticker
3 Red Diamonds per Sticker
3 Red Diamonds per Sticker in Rotated Pyramidal Shape
3 Stacked Red Diamonds in Pyramid Formation per Sticker
4 Red Diamonds in Square Formation per Sticker
Width (Metric) Tape:
55 mm
55.00 mm
75.00 mm
105.00 mm
Material:
Paper
Polypropylene
Unit Size:
Pack of 200
Pack of 300
Pack of 400
This item is not returnable. View return policy

Product Code. 30041477

Brand: Brady™ 622321

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Supplied with pre-printed red diamonds, ready for you to print the required warning, compliant to the latest regulations.

For you to print the required warning, compliant to the latest regulations.

Especificaciones

Certifications/Compliance UL Recognized
Description Pre-printed Desktop Printer Labels for CLP-GHS Hazardous Substances
Adhesive Type Acrylic, Permanent
Coating None
Color Red on White
Finish Gloss
Format Die-cut
Material Polypropylene
Printable Sides Front
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
Display Text 3 Red Diamonds per Sticker in Rotated Pyramidal Shape
For Use With (Application) Hazard communication identification
For Use With (Equipment) BBP11, BBP12, PR Plus, THT 25 mm Core Printers
Labels per Row 1
Min. Operating Temperature (Metric) -20°C
Model R6000HF
No. per Pack 300 Labels/Roll
Quantity 300 Stickers
Shape Rectangle
Size 75.00 mm (W) x 110.00 mm (H)
Temperature Range (English) -4°F to 212°F
Temperature Range (Metric) -20°C to +100°C
Thickness (English) 0.0031 in.
Thickness (Metric) 0.079 mm
Dimensions (LxW) 75.00 mm (W) x 110.00 mm (H)
Length (English) 4.33 in.
Length (Metric) 110.00 mm
Width (English) 2.95 in.
Width (Metric) 75.00 mm
Width (English) Printable Area 75.00 mm
Width (Metric) Tape 75.00 mm
Height (English) 4.33 in.
Height (Metric) 110.00 mm
Alternate Part Number THTCLP-M-L3A-7594-0.3-SC
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado