missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SMAGP Polyclonal Antibody

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5148784

Product Code. 30535975

  • £415.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

Small cell adhesion glycoprotein (SMAGP) is a gene located on chromosome 1p33 that encodes a transmembrane protein involved in cell adhesion processes. SMAGP is primarily expressed in epithelial tissues and plays a role in maintaining cellular architecture and tissue integrity. This glycoprotein contributes to the organization of cell-cell junctions, which are critical for epithelial barrier function and communication between cells. Altered expression of SMAGP has been observed in various cancers, including gastric and colorectal cancers, where it may influence tumor progression and metastasis by affecting cellular adhesion and epithelial-to-mesenchymal transition (EMT). Research into SMAGP continues to explore its functional mechanisms in epithelial biology and its potential as a biomarker for cancer diagnosis and prognosis. By providing insights into the molecular pathways governing cell adhesion and tumorigenesis, studies on SMAGP contribute to the broader understanding of cancer biology and therapeutic opportunities.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SMAGP
Polyclonal
Unconjugated
SMAGP
hSMAGP; Smagp; Small cell adhesion glycoprotein; small cell transmembrane and glycosylated protein; small transmembrane and glycosylated protein; small trans-membrane and glycosylated protein
Goat
Antigen affinity chromatography
RUO
57228
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
Q0VAQ4
SMAGP
E. coli-derived recombinant human SMAGP, Thr2-Thr34, Phe56-Ile97.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.