missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat SMAGP Antibody, R&D Systems™
Goat Polyclonal Antibody
£146.86 - £408.36
Specifications
Antigen | SMAGP |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18745267
|
R&D Systems
AF3967-SP |
25 μg |
£146.86
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18000162
|
R&D Systems
AF3967 |
100 μg |
£408.36
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMAGP Polyclonal specifically detects SMAGP in Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SMAGP | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Rat | |
hSMAGP, LOC57228, MGC149453, MGC149454, small cell adhesion glycoprotein, small cell transmembrane and glycosylated protein, small trans-membrane and glycosylated protein, Small transmembrane and glycosylated protein | |
SMAGP | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
Q7TPF1 | |
57228 | |
E. coli-derived recombinant rat SMAGP Met1-Thr34, Phe56-Ile97 Accession # Q7TPF1 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects rat SMAGP in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant human SMAGP is observed. |
For Research Use Only