missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad7 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93969-0.1ml
This item is not returnable.
View return policy
Description
Smad7 Polyclonal antibody specifically detects Smad7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Smad7 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| CRCS3, FLJ16482, hSMAD7, MAD homolog 7, MAD homolog 8, MAD, mothers against decapentaplegic homolog 7 (Drosophila), MADH7mothers against decapentaplegic homolog 7, MADH8, Mothers against decapentaplegic homolog 8, Mothers against decapentaplegic, drosophila, homolog of, 7, Mothers against DPP homolog 7, Mothers against DPP homolog 8, SMAD 7, SMAD family member 7MAD (mothers against decapentaplegic, Drosophila) homolog 7, SMAD, mothers against DPP homolog 7, SMAD, mothers against DPP homolog 7 (Drosophila), Smad7 | |
| A synthetic peptide corresponding to a sequence within amino acids 220-420 of human Smad7 (NP_005895.1). KPTADCPDAVPSSAETGGTNYLAPGGLSDSQLLLEPGDRSHWCVVAYWEEKTRVGRLYCVQEPSLDIFYDLPQGNGFCLGQLNSDNKSQLVQKVRSKIGCGIQLTREVDGVWVYNRSSYPIFIKSATLDNPDSRTLLVHKVFPGFSIKAFDYEKAYSLQRPNDHEFMQQPWTGFTVQISFVKGWGQCYTRQFISSCPCWLE | |
| 0.1 mL | |
| Cell Biology, Signal Transduction | |
| 4092 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction