missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMAD6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599401
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SMAD6.
SMAD6 (mothers against decapentaplegic homolog 6) is an antagonist of signaling by TGF-beta (transforming growth factor) type 1 receptor superfamily members; has been shown to inhibit selectively BMP (bone morphogenetic proteins) signaling by competing with the co-SMAD SMAD4 for receptor-activated SMAD1. SMAD6 is an inhibitory SMAD (I-SMAD) or antagonistic SMAD.
Specifications
| SMAD6 | |
| Polyclonal | |
| Unconjugated | |
| SMAD6 | |
| AOVD2; b2b390Clo; HsT17432; MAD homolog 6; mad homolog 7; Madh6; Madh7; Mothers against decapentaplegic homolog 6; mothers against decapentaplegic, drosophila, homolog of, 6; mothers against DPP homolog 6; Msmad6; SMAD; SMAD 6; SMAD family member 6; SMAD SMAD; SMAD, mothers against DPP homolog 6; SMAD6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17130, 4091 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35182, O43541 | |
| SMAD6 | |
| A synthesized peptide derived from human SMAD6(Accession O43541), corresponding to amino acid residues Q352-A402. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction