missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMAD6 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56440CL1
This item is not returnable.
View return policy
Description
SMAD6 Polyclonal antibody specifically detects SMAD6 in Human,Monkey,Mouse,Primate,Rat,Sheep samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Simple Western,Immunocytochemistry/Immunofluorescence
Specifications
| SMAD6 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Monkey, Mouse, Primate, Rat | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Simple Western, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| hSMAD6, HsT17432, MAD homolog 6, MADH6SMAD, mothers against DPP homolog 6 (Drosophila), MADH7, mothers against decapentaplegic homolog 6, Mothers against decapentaplegic, drosophila, homolog of, 6, Mothers against DPP homolog 6, SMAD 6, SMAD family member 6MAD, mothers against decapentaplegic homolog 6 (Drosophila), SMAD, mothers against DPP homolog 6, Smad6 | |
| Synthetic peptide made to an internal portion of human SMAD6 (between amino acids 30-150) [UniProt O43541]. | |
| 0.1 mL | |
| Stem Cell Signaling Pathway | |
| 4091 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction