missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad5, p Ser463, p Ser465 Antibody (3A9), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26390-100ul
This item is not returnable.
View return policy
Description
Smad5 Monoclonal antibody specifically detects Smad5 in Human samples. It is validated for Western Blot, ELISA
Specifications
| Smad5 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 3A9 | |
| Western Blot 1:500-1:5000, ELISA | |
| hSmad5, JV5-1DKFZp781O1323, MAD homolog 5, MAD, mothers against decapentaplegic homolog 5 (Drosophila), MADH5mothers against decapentaplegic homolog 5, mothers against decapentaplegic homolog 5, mothers against decapentaplegic, drosophila, homolog of, 5, Mothers against DPP homolog 5, SMA- and MAD-related protein 5, SMAD 5, SMAD family member 5DKFZp781C1895, SMAD, mothers against DPP homolog 5, SMAD, mothers against DPP homolog 5 (Drosophila), Smad5 | |
| A synthesized peptide derived from Human SMAD5 [p Ser463, p Ser465] [UniProt Q99717] | |
| 100 μL | |
| Phospho Specific, Signal Transduction | |
| 4090 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction