Learn More
SMAD5 Mouse anti-Human, Unconjugated, Clone: 1A6A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67052-1-IG-20UL
Description
The SMAD5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SMAD5. This antibody recognizes human, mouse, rat antigen. The SMAD5 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| SMAD5 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry | |
| 1A6A4 | |
| Unconjugated | |
| 20 μL | |
| 4090 | |
| IgG2a |