missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMAD4 Recombinant Rabbit Monoclonal Antibody (PD01-37)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550750
This item is not returnable.
View return policy
Description
Positive control: PC-12 cell lysate, SH-SY5Y cell lysate, human lung carcinoma tissue, human testis tissue, human stomach carcinoma tissue, PANC-1. Predicted band size: 60 kDa Subcellular Location: Cytoplasm, Nucleus.
Smad4 is encoded by the Smad4 gene that is located on chromosome 18 in humans. Smad4 is also known as Smad family member 4, Sma- and Mad-relatedprotein 4 or Mothers against decapentaplegic (MAD) homolog 4. Smad4 contains N-terminal MH1 (MAD homology 1) and C-terminal MH2 (MAD homology 2) globular domains that are involved in DNA binding and protein interactions respectively. Binding of the TGF-beta superfamily of ligands that includes Transforming Growth Factor -beta (TGF-beta) and bone morphogenetic protein (BMP) to its cognate receptor allows phosphorylation of Smad 1, 2, 3, 5 and 8 (R-Smad, Receptor Smad). This signals for heterotrimerization with Smad4 (co-Smad, co-mediator Smad) and translocation of the complex to the nucleus. Inhibitory or antagonistic Smad (I-Smad) that includes Smad 6 and 7, interact with activated R-Smads and attenuate the signaling pathway. Smad4 acts as a tumor suppressor protein by transcriptionally regulating its target genes such as Cyclin D1 (downregulation) and collagen (upregulation) that inhibit cell proliferation. Dephosphorylation regulates nuclear export and nucleocytoplasmic dynamics of Smads.
Specifications
| SMAD4 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O70437, P97471, Q13485 | |
| SMAD4 | |
| Synthetic peptide within Human Smad4 aa 500-552/552 (C terminal). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| PD01-37 | |
| Unconjugated | |
| SMAD4 | |
| AW743858; D18Wsu70e; Deleted in Pancreatic Carcinoma 4 (DPC4); deleted in pancreatic carcinoma locus 4; deletion target in pancreatic carcinoma 4; Deletion target in pancreatic carcinoma 4 homolog; DPC4; hSMAD4; JIP; MAD (mothers against decapentaplegic Drosophila) homolog 4; MAD homolog 4; MAD homolog 4 (MADH4); Madh4; mothers against decapentaplegic homolog 4; Mothers against decapentaplegic homolog 4 (MADH4); mothers against decapentaplegic, Drosophila, homolog of, 4; mothers against DPP homolog 4; MYHRS; SMAD; SMAD 4; SMAD family member 4; SMAD, mothers against DPP homolog 4; SMAD4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 17128, 4089, 50554 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction