Learn More
Invitrogen™ SMAD4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596378
Description
Immunogen sequence: PEYWCSIAYF EMDVQVGETF KVPSSCPIVT VDGYVDPSGG DRFCLGQLSN VHRTEAIERA RLHIGKGVQL ECKGEGDVWV RCLSDHAVFV QSYYLDREAG RAPGDAVHKI YPSAYIKVFD LRQCHRQMQQ QAATAQAAAA AQAAAVAGNI PGPGSVGGIA PAISLSAAAG IGVDDLRRLC ILRMSFVKGW GPDYPRQSIK ETPCWIEIHL HRALQLLDEV LHTMPIADPQ PLD Positive Samples: HeLa, SKOV3, A673, 293T, HepG2; Cellular Location: Cytoplasm, Nucleus.
Smad4 is encoded by the Smad4 gene that is located on chromosome 18 in humans. Smad4 is also known as Smad family member 4, Sma- and Mad-relatedprotein 4 or Mothers against decapentaplegic (MAD) homolog 4. Smad4 contains N-terminal MH1 (MAD homology 1) and C-terminal MH2 (MAD homology 2) globular domains that are involved in DNA binding and protein interactions respectively. Binding of the TGF-beta superfamily of ligands that includes Transforming Growth Factor -beta (TGF-beta) and bone morphogenetic protein (BMP) to its cognate receptor allows phosphorylation of Smad 1, 2, 3, 5 and 8 (R-Smad, Receptor Smad). This signals for heterotrimerization with Smad4 (co-Smad, co-mediator Smad) and translocation of the complex to the nucleus. Inhibitory or antagonistic Smad (I-Smad) that includes Smad 6 and 7, interact with activated R-Smads and attenuate the signaling pathway. Smad4 acts as a tumor suppressor protein by transcriptionally regulating its target genes such as Cyclin D1 (downregulation) and collagen (upregulation) that inhibit cell proliferation. Dephosphorylation regulates nuclear export and nucleocytoplasmic dynamics of Smads.
Specifications
| SMAD4 | |
| Polyclonal | |
| Unconjugated | |
| SMAD4 | |
| AW743858; D18Wsu70e; Deleted in Pancreatic Carcinoma 4 (DPC4); deleted in pancreatic carcinoma locus 4; deletion target in pancreatic carcinoma 4; Deletion target in pancreatic carcinoma 4 homolog; DPC4; hSMAD4; JIP; MAD (mothers against decapentaplegic Drosophila) homolog 4; MAD homolog 4; MAD homolog 4 (MADH4); Madh4; mothers against decapentaplegic homolog 4; Mothers against decapentaplegic homolog 4 (MADH4); mothers against decapentaplegic, Drosophila, homolog of, 4; mothers against DPP homolog 4; MYHRS; SMAD; SMAD 4; SMAD family member 4; SMAD, mothers against DPP homolog 4; SMAD4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17128, 4089, 50554 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O70437, P97471, Q13485 | |
| SMAD4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 320-552 of human Smad4 (NP_0053501). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.