missing translation for 'onlineSavingsMsg'
Learn More

Smad4 Antibody (SMAD/6309R), FITC, Novus Biologicals™

Product Code. 18325232
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18325232 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18325232 Supplier Bio-Techne Supplier No. NBP314116F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Smad4 Monoclonal antibody specifically detects Smad4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Smad4
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SMAD/6309R
Conjugate FITC
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias DPC4SMAD, mothers against DPP homolog 4 (Drosophila), hSMAD4, JIP, MAD homolog 4, MADH4mothers against decapentaplegic homolog 4, mothers against decapentaplegic homolog 4, mothers against decapentaplegic, Drosophila, homolog of, 4, SMAD 4, SMAD family member 4MAD, mothers against decapentaplegic homolog 4 (Drosophila), SMAD, mothers against DPP homolog 4, Smad4
Host Species Rabbit
Immunogen Recombinant human full-length (aa1-552) Smad4 protein (Uniprot: Q13485 )
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4089
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.