missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad3, p Ser425, p Ser423 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Smad3 Polyclonal antibody specifically detects Smad3 in Human, Mouse samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | Smad3 |
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DKFZp586N0721, DKFZp686J10186, hMAD-3, hSMAD3, HsT17436, JV15-2MGC60396, MAD homolog 3, mad protein homolog, MAD, mothers against decapentaplegic homolog 3, MAD, mothers against decapentaplegic homolog 3 (Drosophila), mad3, MADH3mad homolog JV15-2, mothers against decapentaplegic homolog 3, Mothers against DPP homolog 3, SMA- and MAD-related protein 3, SMAD 3, SMAD family member 3HSPC193, SMAD, mothers against DPP homolog 3, SMAD, mothers against DPP homolog 3 (Drosophila), Smad3 |
| Host Species | Rabbit |
| Immunogen | Smad3 [p Ser423, p Ser425] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a dual phosphorylated synthetic peptide corresponding to a c-terminal region with Serine 423 and Serine 425 of human Smad3 protein. (Uniprot: P84022) |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?