missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMAD3 Mouse anti-Human, Mouse, Clone: 1F10B2, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | SMAD3 |
|---|---|
| Clone | 1F10B2 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16848465
|
Proteintech
66516-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16838465
|
Proteintech
66516-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMAD3 belongs to the SMAD family of proteins. They are similar to the gene products of the Drosophila gene ′mothers against decapentaplegic′ (Mad) and the C. elegans gene Sma. SMAD proteins are signal transducers and transcriptional modulators that mediate multiple signaling pathways. This protein functions as a transcriptional modulator activated by transforming growth factor-beta and is thought to play a role in the regulation of carcinogenesis.Specifications
| SMAD3 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| hMAD 3, hSMAD3, HSPC193, HsT17436, JV15 2, MAD homolog 3, Mad3, MADH3, Mothers against DPP homolog 3, SMAD 3, SMAD family member 3, SMAD3 | |
| Smad3 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1F10B2 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Mouse, Human | |
| P84022, Q8BUN5 | |
| 17127, 4088 | |
| Peptide | |
| Primary | |
| -20°C | |
| SMAD3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title