missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMAD2(phospho S467) & SMAD2 Protein Phosphorylation Antibody Pair, Abnova™
Antibody Pair
Brand: Abnova DP0240.1Set
This item is not returnable.
View return policy
Description
This protein phosphorylation antibody pair set comes with two antibodies, one against the SMAD2 protein, and the other against the specific S467 phosphorylated site of SMAD2 for use in in situ Proximity Ligation Assay.Specifications
| SMAD2 | |
| Unconjugated | |
| JV18/JV18-1/MADH2/MADR2/MGC22139/MGC34440/hMAD-2/hSMAD2 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Phospho-SMAD2 S467 rabbit polyclonal antibody (20 ul) In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. SMAD2 mouse monoclonal antibody (40 ug) In 1x PBS, pH 7.2 *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| In situ PLA | |
| SMAD family member 2 | |
| SMAD2 | |
| RUO | |
| 4087 | |
| Human | |
| Ab Pair |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction