missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Smad2 Polyclonal antibody specifically detects Smad2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Smad2 |
| Applications | Western Blot |
| Host Species | Rabbit |
| Quantity | 0.1 mL |
| Research Discipline | Cancer, Cell Cycle and Replication, Metastasis, Phospho Specific, Stem Cell Signaling Pathway, Wnt Signaling Pathway |
| Content And Storage | Store at 4C in the dark. |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?