missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMAD1, Mouse anti-Human, Clone: 2E9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00004086-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: MNVTSLFSFTSPAVKRLLGWKQGDEEEKWAEKAVDALVKKLKKKKGAMEELEKALSCPGQPSNCVTIPRSLDGRLQVSHRKGLPHVIYCRVWRWPDLQSHHELKPLECCEFPFGSKQKEVCINPYHYKRVESPVLPPVLVPRHSEYNPQHSLLAQFRNLGQNEPHMPLNATFPDSFQQPNSHPFPHSPNSSYPNSPGSSSSTYPHSPTSSDPGSPFQMPADTPPPAYLPPEDPMTQDGSQPMDTNMMAPPLPSEINRGDVQAVAYEEPKHWCSIVYYELNNRVGEAFHASSTSVLVDGFTDPSNNKNRFCLGLLSNVNRNSTIENTRRHIGKGVHLYYVGGEVYAECLSDSSIFVQSRNCNYHHGFHPTTVCKIPSGCSLKIFNNQEFAQLLAQSVNHGFETVYELTKMCTIRMSFVKGWGAEYHRQDVTSTPCWIEIHLHGPLQWLDKVLTQMGSPHNPISSVSSpecifications
| SMAD1 | |
| Monoclonal | |
| Unconjugated | |
| SMAD family member 1 | |
| BSP1/JV4-1/JV41/MADH1/MADR1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 2E9 | |
| In 1x PBS, pH 7.4 | |
| BC001878 | |
| SMAD1 | |
| SMAD1 (AAH01878.1, 1 a.a. ∼ 465 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 4086 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction