missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMAC/Diablo Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56165
This item is not returnable.
View return policy
Description
SMAC/Diablo Polyclonal specifically detects SMAC/Diablo in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| SMAC/Diablo | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Simple Western 1:400, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000, Immunohistochemistry-Frozen | |
| 0610041G12Rik, diablo, IAP-binding mitochondrial protein, Direct IAP-binding protein with low pI, mitochondrial Smac protein, Second mitochondria-derived activator of caspase, Smac, SMACmitochondrial | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| Smac | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Whole antisera with 0.05% Sodium Azide | |
| DIABLO | |
| Full-length recombinant human SMAC was used as immunogen. | |
| 0.05 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 56616 | |
| Human, Mouse, Rat | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction