missing translation for 'onlineSavingsMsg'
Learn More

SMAC/Diablo Antibody (78-1-118), Novus Biologicals™

Product Code. 18221201 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
Unit Size:
0.05mL
0.2mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18221201 0.05 mL 0.05mL
18159919 0.2 mL 0.2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18221201 Supplier Novus Biologicals Supplier No. NB500213SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SMAC/Diablo Monoclonal specifically detects SMAC/Diablo in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMAC/Diablo
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 78-1-118
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10 - 1:500
Formulation Ascites with 0.1% Sodium Azide
Gene Accession No. Q9NR28
Gene Alias 0610041G12Rik, diablo, IAP-binding mitochondrial protein, Direct IAP-binding protein with low pI, mitochondrial Smac protein, Second mitochondria-derived activator of caspase, Smac, SMACmitochondrial
Gene Symbols DIABLO
Host Species Mouse
Immunogen Recombinant mature human Smac (lacks amino acids 1-55 present in Smac precursor). [UniProt# Q9NR28]
Molecular Weight of Antigen 25 kDa
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Mitochondrial Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 56616
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG3, IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.