missing translation for 'onlineSavingsMsg'
Learn More

SMAC/Diablo Antibody (006), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18353181 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18353181 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18353181 Supplier Novus Biologicals Supplier No. NBP289251JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SMAC/Diablo Monoclonal specifically detects SMAC/Diablo in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Especificaciones

Antigen SMAC/Diablo
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 6
Conjugate Janelia Fluor 646
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM Sodium Borate
Gene Alias 0610041G12Rik, diablo, IAP-binding mitochondrial protein, Direct IAP-binding protein with low pI, mitochondrial Smac protein, Second mitochondria-derived activator of caspase, Smac, SMACmitochondrial
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human SMAC/Diablo (NP_063940.1; Ala 56-Asp 239).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cellular Markers, Mitochondrial Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 56616
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.