missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLURP1 Monoclonal Antibody (569317)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524256
This item is not returnable.
View return policy
Description
In direct ELISAs, no cross-reactivity with recombinant mouse SLURP1 is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Specifications
| SLURP1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P55000 | |
| SLURP1 | |
| E. coli-derived recombinant human SLURP1 Ala22-Leu103. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin) | |
| 569317 | |
| Unconjugated | |
| SLURP1 | |
| anti-neoplastic urinary protein; ANUP; ARS; ARS component B; ARS(component B)-81/S; ArsB; LY6LS; lymphocyte antigen 6-like secreted; MDM; secreted LY6/PLAUR domain containing 1; secreted Ly6/uPAR related protein 1; secreted Ly-6/uPAR-related protein 1; SLURP1; SLURP-1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 57152 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction