missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLUG Recombinant Rabbit Monoclonal Antibody (BLR139H)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544466
This item is not returnable.
View return policy
Description
SLUG Recombinant Monoclonal Antibody for Western Blot, IP
Slug, a member of the Snail family of C2H2-type zinc finger transcription factors, was initially identified to be involved in epithelial-mesenchymal transitions as well as the formation of the neural tube during vertebrate embryogenesis. Like Snail, Slug transcription can be induced by growth factors such as FGF, BMP, and TGF-beta. Once expressed, Slug will bind E-box regions of promoters and repress transcription of genes such as E-cadherin and Claudin-1. More recently, its expression in breast, esophogeal, and colorectal carcinomas has been correlated with poor prognosis for survival. Furthermore, Slug can protect hemopoietic progenitor cells from radiation-induced apoptosis by repressing the p53-mediated transcription of Puma, a BH3-only antagonist of the anti-apoptotic members of the Bcl-2 family.
Specifications
| SLUG | |
| Recombinant Monoclonal | |
| 0.4 mg/mL | |
| BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6 | |
| O43623 | |
| SNAI2 | |
| Peptide representing a region between residue 75 and 125 of Human Zinc finger protein SNAI2 (aka Slug). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| BLR139H | |
| Unconjugated | |
| SNAI2 | |
| MGC10182; neural crest transcription factor Slug; Protein snail homolog 2; SLUG; slug (chicken homolog), zinc finger protein; Slug, chicken homolog; Slugh; SLUGH1; Snai2; snail family transcriptional repressor 2; snail family zinc finger 2; snail homolog 2; Snail2; snail-like protein 2; WS2D; Zinc finger protein SNAI2; zinc finger transcription factor snail 2 | |
| Rabbit | |
| purified | |
| RUO | |
| 6591 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction