missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Slug Antibody (OTI1A6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74229CL1
This item is not returnable.
View return policy
Description
Slug Monoclonal antibody specifically detects Slug in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),CyTOF
Specifications
| Slug | |
| Monoclonal | |
| CoraFluor 1 | |
| MGC10182, Neural crest transcription factor Slug, Protein snail homolog 2, slug homolog, zinc finger protein (chicken), SLUGH, SLUGzinc finger protein, snail 2, snail homolog 2 (Drosophila), WS2D, zinc finger protein SNAI2 | |
| Full length human recombinant protein of human SNAI2 (NP_003059) produced in E.coli. | |
| 0.1 mL | |
| Neuronal Cell Markers, Phospho Specific | |
| 6591 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), CyTOF | |
| OTI1A6 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction