missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Slug Antibody (666633) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7408JF525
This item is not returnable.
View return policy
Description
Slug Monoclonal antibody specifically detects Slug in Human samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| Slug | |
| Monoclonal | |
| Janelia Fluor 525 | |
| MGC10182, Neural crest transcription factor Slug, Protein snail homolog 2, slug homolog, zinc finger protein (chicken), SLUGH, SLUGzinc finger protein, snail 2, snail homolog 2 (Drosophila), WS2D, zinc finger protein SNAI2 | |
| E. coli-derived recombinant human Slug, Met1-His268, Accession # O43623 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 666633 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Neuronal Cell Markers, Phospho Specific | |
| 6591 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction