missing translation for 'onlineSavingsMsg'
Learn More

Whirl-Pak™ Sludge Judge II™ Components

Product Code. p-9012007 Shop All Cole-Parmer Products
Click to view available options
Type:
Extension
Includes:
Middle Section
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15362694

Brand: Whirl-Pak™ 0546532

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Separate extension section replacement parts for the Sludge Judge II.

Replacement parts for Sludge Judge™ II. Complete sampler unit available under part number B01355

Specifications

Certifications/Compliance ISO 9001 and ISO 13485
Volume (Metric) 177 mL/ft.
For Use With (Application) Sludge Judge II
For Use With (Equipment) Sludge Judge II
Graduated Yes
Includes Middle Section
Type Extension
Length (English) 5 ft.
Length (Metric) 1.53 m
Model B01357
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.