Learn More
Invitrogen™ SLU7 Monoclonal Antibody (P262)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518296
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, human SLU7-transfected 293T. Predicted reactivity: Human (99%), Mouse (98%), Rat (97%), Zebrafish (80%), Chicken (86%), Rhesus Monkey (99%), Chimpanzee (99%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Pre-mRNA splicing occurs in two sequential transesterification steps. The protein encoded by this gene is a splicing factor that has been found to be essential during the second catalytic step in the pre-mRNA splicing process. It associates with the spliceosome and contains a zinc knuckle motif that is found in other splicing factors and is involved in protein-nucleic acid and protein-protein interactions.
Specifications
| SLU7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O95391 | |
| SLU7 | |
| Recombinant protein encompassing a sequence within the center region of human SLU7. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| P262 | |
| Unconjugated | |
| SLU7 | |
| 9G8; AU018913; D11Ertd730e; D3Bwg0878e; hSlu7; MGC9280; Pre-mRNA-splicing factor SLU7; SLU7; SLU7 homolog, splicing factor; SLU7 splicing factor homolog; SLU7 splicing factor homolog (S. cerevisiae); splicing factor; step II splicing factor SLU7; zinc knuckle motif containing | |
| Mouse | |
| Protein G | |
| RUO | |
| 10569 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.