missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLU7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-20403
This item is not returnable.
View return policy
Description
SLU7 Polyclonal specifically detects SLU7 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SLU7 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal | |
| hSlu7, MGC9280, pre-mRNA-splicing factor SLU7,9G8, SLU7 splicing factor homolog (S. cerevisiae), splicing factor, step II splicing factor SLU7, zinc knuckle motif containing | |
| Rabbit | |
| 68 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse. Species reactivity based on sequence identity: Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| O95391 | |
| SLU7 | |
| Recombinant protein encompassing a sequence within the center region of human SLU7. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 10569 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction