missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ SLU-PP-332
Description
SLU-PP-332 is a potent and selective pan estrogen related receptor (ERR) agonist (EC50 values are 98 nM, 230 nM and 430 nM for ERRα, ERRβ and ERRγ respectively). In a mouse model of metabolic syndrome SLU-PP-332 increases energy expenditure and fatty acid oxidation, decreases fat mass, and improves insulin sensitivity. Also increases mitochondrial respiration in vitro.
Specifications
Specifications
| Chemical Name or Material | (E)-4-Hydroxy-N'-(naphthalen-2-ylmethylene)benzohydrazide |
| CAS | 303760-60-3 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Target | Estrogen and Related Receptor Agonists |
| Molecular Formula | C18H14N2O2 |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?