missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse SLPI Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0503
Code : NEW
Additional Details : Weight : 0.01000kg
Description
SLPI Synthetic Peptide, PEP-0503, from Invitrogen.
This peptide corresponds to 17 amino acids near the center of mouse SLPI.PEP-0503 can be used as a blocking peptide with polyclonal antibody PA5-20386.Specifications
20568 | |
purified | |
Mouse SLPI | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Mouse | |
P97430 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
Slpi | |
0.02% sodium azide | |
50 μg | |
PBS |