missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLPI Rabbit anti-Human, Clone: 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89438-100UL
This item is not returnable.
View return policy
Description
SLPI Monoclonal specifically detects SLPI in Human samples. It is validated for ELISA.
Specifications
| SLPI | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 1 | |
| ELISA 1:5000-1:10000 | |
| ALK1, ALPProtease inhibitor WAP4, antileukoproteinase, BLPISeminal proteinase inhibitor, HUSI, HUSI-I, Mucus proteinase inhibitor, secretory leukocyte peptidase inhibitor, Secretory leukocyte protease inhibitor, secretory leukocyte protease inhibitor (antileukoproteinase), WAP four-disulfide core domain protein 4, WAP4HUSI-1, WFDC4MPI | |
| Recombinant Human SLPI Protein | |
| 100 μg | |
| Cancer | |
| 6590 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction