missing translation for 'onlineSavingsMsg'
Learn More

SLPI Antibody, mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18419652 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419652 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419652 Supplier Novus Biologicals Supplier No. NBP176803MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLPI Polyclonal antibody specifically detects SLPI in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLPI
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ALK1, ALPProtease inhibitor WAP4, antileukoproteinase, BLPISeminal proteinase inhibitor, HUSI, HUSI-I, Mucus proteinase inhibitor, secretory leukocyte peptidase inhibitor, Secretory leukocyte protease inhibitor, secretory leukocyte protease inhibitor (antileukoproteinase), WAP four-disulfide core domain protein 4, WAP4HUSI-1, WFDC4MPI
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide from near the center of human SLPI. The immunogen is located within amino acids 60 - 110 of SLPI. Amino Acid Squence: CLDPVDTPNPTRRKPGK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6590
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.