missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLPI Antibody (001), CoraFluor™ 1, Novus Biologicals™
Description
SLPI Monoclonal antibody specifically detects SLPI in Human samples. It is validated for ELISA,Immunocytochemistry/Immunofluorescence
Specifications
Specifications
| Antigen | SLPI |
| Applications | ELISA, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Clone | 001 |
| Conjugate | CoraFluor 1 |
| Formulation | PBS |
| Gene Alias | ALK1, ALPProtease inhibitor WAP4, antileukoproteinase, BLPISeminal proteinase inhibitor, HUSI, HUSI-I, Mucus proteinase inhibitor, secretory leukocyte peptidase inhibitor, Secretory leukocyte protease inhibitor, secretory leukocyte protease inhibitor (antileukoproteinase), WAP four-disulfide core domain protein 4, WAP4HUSI-1, WFDC4MPI |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human SLPI (Uniprot#: P03973; Met1-Ala132). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?