missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLP-76/LCP2 Antibody (SLP-76/03), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-537CL1
This item is not returnable.
View return policy
Description
SLP-76/LCP2 Monoclonal antibody specifically detects SLP-76/LCP2 in Human,Mouse,Porcine samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| SLP-76/LCP2 | |
| Monoclonal | |
| CoraFluor 1 | |
| lymphocyte cytosolic protein 2, lymphocyte cytosolic protein 2 (SH2 domain containing leukocyte protein of76kDa), lymphocyte cytosolic protein 2 (SH2 domain-containing leukocyte protein of76kD), SH2 domain-containing leukocyte protein of 76 kDa, SH2 domain-containing leukocyte protein of 76kD, SLP-76, SLP-76 tyrosine phosphoprotein, SLP7676 kDa tyrosine phosphoprotein | |
| Bacterially expressed fusion protein representing amino acids 216-434 of human SLP76 with histidine tag | |
| 0.1 mL | |
| Phospho Specific, Signal Transduction | |
| 3937 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| SLP-76/03 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction