missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLP-2 Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NB100-75482
This item is not returnable.
View return policy
Description
SLP-2 Polyclonal specifically detects SLP-2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| SLP-2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| EPB72-like protein 2, HSPC108, SLP2, SLP-2stomatin-like protein 2, stomatin (EPB72)-like 2, stomatin-like 2 | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| Primary | |
| STOML2 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1.0 mg/mL | |
| Western Blot | |
| Q53G29 | |
| STOML2 | |
| The immunogen for this antibody was a sequence from 171a.a. to 356a.a. from STOML2(a 356a.a. long protein) | |
| 0.05 mg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 30968 | |
| Human, Mouse, Rat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction