missing translation for 'onlineSavingsMsg'
Learn More

Slow Skeletal Muscle Myosin Heavy Chain Antibody (96J), Novus Biologicals™

Mouse Monoclonal Antibody

£159.00 - £310.00

Specifications

Antigen Slow Skeletal Muscle Myosin Heavy Chain
Clone 96J
Concentration 1 mg/mL
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Applications Western Blot, Immunohistochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18692748
View Documents
Novus Biologicals
NBP2-50298-0.025ml
0.025 mL
£159.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18682938
View Documents
Novus Biologicals
NBP2-50298
0.1 mL
£310.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Slow Skeletal Muscle Myosin Heavy Chain Monoclonal antibody specifically detects Slow Skeletal Muscle Myosin Heavy Chain in Human, Mouse, Rat, Chicken, Rabbit samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

Slow Skeletal Muscle Myosin Heavy Chain
1 mg/mL
Western Blot, Immunohistochemistry
Unconjugated
Mouse
Cardiovascular Biology, Cellular Signaling, Developmental Biology, Stem Cells
Ascites
IgG1
Ascites
96J
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Monoclonal
Ascites
RUO
Human, Mouse, Rat, Chicken, Rabbit
Purified human skeletal muscle myosin
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.