missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Slo3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05611-100ul
This item is not returnable.
View return policy
Description
Slo3 Polyclonal antibody specifically detects Slo3 in Rat samples. It is validated for Western Blot
Specifications
| Slo3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Calcium-activated potassium channel subunit alpha-3, KCa5, KCa5.1, Kcnma3, KCNMC1Calcium-activated potassium channel, subfamily M subunit alpha-3, potassium channel subfamily U member 1, potassium channel, subfamily U, member 1, Slo3, Slowpoke homolog 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 920-1149 of human Slo3 (NP_001027006.2). LELLQMLVTGGVSSQLEQHLDKDKVYGVADSCTSLLSGRNRCKLGLLSLHETILSDVNPRNTFGQLFCGSLDLFGILCVGLYRIIDEEELNPENKRFVITRPANEFKLLPSDLVFCAIPFSTACYKRNEEFSLQKSYEIVNKASQTTETHSDTNCPPTIDSVTETLYSPVYSYQPRTNSLSFPKQIAWNQSRTNSIISSQIPLGDNAKENERKTSDEVYDEDPFAYSEPL | |
| 100 μg | |
| Cell Biology, Neuroscience | |
| 157855 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction