missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122311
This item is not returnable.
View return policy
Description
SLN, also named as Sarcolipin, belongs to the sarcolipin family. It is associated with calcium ATPase SERCA1. It inhibits SERCA pumps. SLN, a key regulator of cardiac sarco (endo)plasmic reticulum (SR) Ca(2+) ATPase, is predominantly expressed in atria and mediates beta-adrenergic responses.
Specifications
| SLN | |
| Polyclonal | |
| Unconjugated | |
| Sln | |
| 2310045A07Rik; Sarcolipin; SLN | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 6588 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O00631 | |
| Sln | |
| Met1-Tyr31 (Accession O00631), with N-terminal His and GST Tag. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction