Learn More
Invitrogen™ SLMAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551708
Description
Immunogen sequence: SEYEKEITSL QNSFQLRCQQ CEDQQREEAT RLQGELEKLR KEWNALETEC HSLKRENVLL SSELQRQEKE LHNSQKQSLE LTSDLSILQM SRKELENQVG SLKEQHLRDS ADLKTLLSKA ENQAKDVQKE YEKTQTVLSE LKLKFEMTEQ Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.
Sarcolemmal membrane associated protein (SLMAP) is a tail-anchored membrane protein involved in the docking and membrane fusion process via protein-protein interactions. Deregulation of SLMAP expression plays an important role in type 2 diabetes, and changes in SLMAP expression are linked to hyperglycemia and diabetes. SLMAPs are components of cardiac membranes and have been identified as a novel regulator of cardiac function. There are several isoforms of SLMAPs with varying molecular size (35, 45, 63, and 83-91 kDa) as a result of alternative splicing. The short 35- kDa isoform is predominantly expressed in cardiac and slow-twitch skeletal muscle, while the 83- to 91- kDa isoform is ubiquitous.
Specifications
| SLMAP | |
| Polyclonal | |
| Unconjugated | |
| Slmap | |
| D330001L02Rik; KIAA1601; mKIAA1601; sarcolemma associated protein; sarcolemmal membrane-associated protein; Sarcolemmal-associated protein; Slap; Slmap; UNQ1847/PRO3577 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 290533, 7871 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P0C219, Q14BN4 | |
| Slmap | |
| Recombinant protein corresponding to Human SLMAP. Recombinant protein control fragment (Product #RP-102135). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.