missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: TTDAQMDEQD LNEPLAKVSL LKALLEEERK AYRNQVEEST KQIQVLQAQL QRLHIDTENL REEKDSEITS TRDELL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.
Sarcolemmal membrane associated protein (SLMAP) is a tail-anchored membrane protein involved in the docking and membrane fusion process via protein-protein interactions. Deregulation of SLMAP expression plays an important role in type 2 diabetes, and changes in SLMAP expression are linked to hyperglycemia and diabetes. SLMAPs are components of cardiac membranes and have been identified as a novel regulator of cardiac function. There are several isoforms of SLMAPs with varying molecular size (35, 45, 63, and 83-91 kDa) as a result of alternative splicing. The short 35- kDa isoform is predominantly expressed in cardiac and slow-twitch skeletal muscle, while the 83- to 91- kDa isoform is ubiquitous.
Specifications
Specifications
| Antigen | SLMAP |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Slmap |
| Gene Accession No. | P0C219, Q14BN4, Q3URD3 |
| Gene Alias | D330001L02Rik; KIAA1601; mKIAA1601; sarcolemma associated protein; sarcolemmal membrane-associated protein; Sarcolemmal-associated protein; Slap; Slmap; UNQ1847/PRO3577 |
| Gene Symbols | Slmap |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
