missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLMAP Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-06131CL1
This item is not returnable.
View return policy
Description
SLMAP Polyclonal antibody specifically detects SLMAP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SLMAP | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 7871 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| FLJ42206, KIAA1601Sarcolemmal-associated protein, MGC138760, sarcolemma associated protein, sarcolemmal membrane-associated protein, SLAPMGC138761 | |
| Produced in rabbits immunized with E. coli-derived Human SLMAP fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido