missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLK Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19743-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody recognizes the C-term of SLK.
SLK mediates apoptosis and actin stress fiber dissolution.Specifications
| SLK | |
| Polyclonal | |
| Unconjugated | |
| SLK | |
| CTCL tumor antigen se20 9, hSLK, KIAA0204, LOSK, se20 9, SLK, STE20 like kinase, STE20 like kinase (yeast), STE20 related kinase, STK2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20874, 9748 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O54988, Q9H2G2 | |
| SLK | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction