missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549878
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total SLK protein.
SLK Polyclonal antibody specifically detects SLK in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Specifications
| SLK | |
| Polyclonal | |
| Unconjugated | |
| SLK | |
| 9A2; AV021402; AW411554; bA16H23.1; CTCL tumor antigen se20-9; Etk4; hSLK; Kiaa0204; Long Ste20-like Kinase; LOSK; mKIAA0204; mSLK; se20-9; serine/threonine kinase 2; serine/threonine-protein kinase 2; Sk2; Slk; SMAK; SNF1 (sucrose nonfermenting, yeast, homolog)-like kinase, SNF1 sucrose nonfermenting like kinase; SNF1 sucrose nonfermenting like kinase; STE20 like kinase; STE20-like kinase; STE20-like serine/threonine-protein kinase; STE20-related kinase; STE20-related kinase SMAK; Ste20-related serine/threonine kinase; STE20-related serine/threonine-protein kinase; Stk2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20874, 54308, 9748 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O08815, O54988, Q9H2G2 | |
| SLK | |
| Synthesized peptide derived from C-terminal of human SLK. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction