missing translation for 'onlineSavingsMsg'
Learn More

SLK Antibody, DyLight 405, Novus Biologicals™

Product Code. 18378212 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18378212 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18378212 Supplier Novus Biologicals Supplier No. NBP298859V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLK Polyclonal specifically detects SLK in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLK
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias bA16H23.1, CTCL tumor antigen se20-9, EC 2.7.11, EC 2.7.11.1, hSLK, KIAA0204LOSK, Long Ste20-like Kinase, se20-9, Serine/threonine-protein kinase 2, SNF1 (sucrose nonfermenting, yeast, homolog)-like kinase, SNF1 sucrosenonfermenting like kinase, SNF1 (sucrose nonfermenting, yeast, homolog)-like kinase, SNF1 sucrosenonfermenting like kinase (yeast), SNF1 sucrose nonfermenting like kinase, STE20-like kinaseSTE20-like kinase (yeast), STE20-like serine/threonine-protein kinase, STE20-related kinase, Ste20-related serine/threonine kinase, STE20-related serine/threonine-protein kinase, STK2MGC133067
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human SLK.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9748
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.