missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLITRK6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA520479
This item is not returnable.
View return policy
Description
A suggested positive control is SK-N-SH cell lysate. PA5-20479 can be used with blocking peptide PEP-0599.
SLIT and NTRK-like family 6 (Slitrk6) is a member a protein family consisting of six homologous transmembrane proteins (Slitrk1-6) that share two conserved leucine-rich repeat domains in the extracellular domain and have significant homology to Slit, a secreted axonal growth-controlling protein. These proteins are also homologous to trk neurotrophin receptors in their intracellular domains. Expression of Slitrk proteins is highly restricted to neural and brain tumor tissues, but varies within the protein family. Slitrk6 expression has been observed in tissues such as tongue, lung, gastrointestinal tract, and pancreas. Like every other Slitrk protein except Slitrk1, overexpression of Slitrk6 inhibited neurite outgrowth in cultured neurons, suggesting that these proteins are involved in the control of neurite outgrowth. At least two isoforms of Slitrk6 are known to exisit.
Specifications
| SLITRK6 | |
| Polyclonal | |
| Unconjugated | |
| SLITRK6 | |
| 4832410J21Rik; DFNMYP; neuronal transmembrane protein Slitrk6; RP11-272M24.2; SLIT and NTRK like family member 6; SLIT and NTRK-like family, member 6; SLIT and NTRK-like protein 6; slit and trk like gene 6; SLITRK6; Sltk6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 239250, 290467, 84189 | |
| 4°C | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8C110, Q9H5Y7 | |
| SLITRK6 | |
| A 16 amino acid peptide from near the carboxy terminus of human Slitrk6. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction